Learn More
Abnova™ ACTR2 Recombinant Protein
Brand:  Abnova™ H00010097-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16124492
  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH14546.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
69.08
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACTR2
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
10097
ACTR2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
ARP2
ACTR2