Learn More
Activated Charcoal, Norit™, SA2, Honeywell™ Fluka™
Product Code. 15657560 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15657560 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15657560 Supplier Honeywell-Fluka Supplier No. 956815KG
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Norit® SA2, from peat, steam activated, powder
  • Norit is a registered trademark of Norit N.V.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7440-44-0
Color Black
pH 6 - 9
Odor Odorless
Molecular Formula C
MDL Number MFCD00133992
Quantity 5 kg
UN Number NONH for all modes of transport
Beilstein 4360473
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.011
PubChem CID 5462310
ChEBI CHEBI:27594
Formula Weight 12.01g/mol
Chemical Name or Material Activated Charcoal Norit™, Norit SA2