Learn More
Activated Charcoal, Norit™, Norit PK 1-3, Honeywell™ Fluka™
Product Code. 15642280 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15642280 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15642280 Supplier Honeywell-Fluka Supplier No. 22874250G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Norit™ PK 1-3, from peat, steam activated, granular
  • Norit is a registered trademark of Norit N.V.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7440-44-0
Color Black
pH 6 - 9
Odor Odorless
Packaging Plastic bottle
Molecular Formula C
MDL Number MFCD00133992
Quantity 250 g
UN Number NONH for all modes of transport
Beilstein 4360473
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.011
PubChem CID 5462310
ChEBI CHEBI:27594
Formula Weight 12.01g/mol
Grade Norit™ PK 1-3
Physical Form Powder
Chemical Name or Material Activated Charcoal Norit™, From peat steam activated