Learn More
Ansell ActivArmr Electrical Insulating Gloves Class 2, Black, 14 in.
Product Code. p-7228764 Shop All Ansell Products
missing translation for 'orderingAttributeHoverText'
Size:
8
9
10
11
12
missing translation for 'unitSize'
1 pair
Product Code. 17825601
missing translation for 'mfr': Ansell RIG214B080
Only null left
Add to basket
Add to basket
ActivArmr Electrical Insulating Gloves provide the ultimate fit, comfort and performance for electrical workers' safety and protection.
  • Performance Profile comfort
  • Natural Rubber Latex for ultimate durability and flexibility
  • Ergonomically-designed hand at rest shape for less hand fatigue
  • Generous flared cuff allows room for clothing and improves ventilation
  • Case hardened finish provides smoother surface for easy donning and doffing performance and protection
  • Product meets all applicable NFPA, OSHA and CSA standards
  • Environmentally-Friendly
  • Proprietary dipping process
  • ISO 14001 certified manufacturing facility
  • Electrical Glove Class 2 offers arc flash protection Class 2 in accordance with EN 61482-1-2
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) CE Number, Category III, Class 2
Color Black
Hand Specific
Material Natural Latex Rubber
Product Type Electrical Insulating Gloves
Size 8
For Use With (Application) Applications with risk of touch voltage, Electrical Contractors, Hybrid engine manufacturing and repair, Maintenance Repairs and Operations in all industrial environments (MRO), Mass Transit Repair and Maintenance, Power maintenance, repair and transmission, Telecom repair near electrical wires
Glove Cuff Color Black
Glove Cuff Style Beaded/Rolled
Series RIG214B
Length (English) 14 in.
Length (Metric) 360 mm
Voltage 17000/25500