missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ACTC (Human) Recombinant Protein
Human ACTC full-length ORF ( AAH09978.1, 1 a.a. - 377 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000070-P01.10ug
Additional Details : Weight : 0.00010kg
Beschreibung
- Sequence: MCDDEETTALVCDNGSGLVKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHTFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKQEYDEAGPSIVHRKCF
Spezifikation
AAH09978.1 | |
actin, alpha, cardiac muscle 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACTC1 | |
GST |
70 | |
Wheat germ expression system | |
10 μg | |
ACTC, CMD1R, CMH11 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |