Learn More
Sanplatec™ Acrylic Vacuum Desiccator YB-3 Type
Click to view available options
Volume (Metric):
30 L
Unit Size:
Each
Description
- High visibility from all sides and durable material
- Vacuum gauge: for 0 to 0.1 MPa
- Front door with silicone gasket and 5 latches
- 2 valves equipped for vacuum and gas replacement
- Rack shelves can be added or subtracted enabling you to increase or decrease the number of shelves to your needs
- Up to 8 levels can be held in this model
Specifications
Specifications
| Color | Transparent |
| Material | PMMA |
| Product Type | Vacuum Desiccator |
| For Use With (Application) | Dehumidification |
| No. of Shelves | 4 |
| Volume (Metric) | 30 L |
| Dimensions (D x W x H) Interior | 240 x 220 x 460 mm |
| Width (Metric) | 341 mm |
| Height (Metric) | 590 mm |
| Depth (Metric) | 445 mm |
| Show More |