Learn More
Cytiva Acrylic Plastic Thermostat Jacket
Product Code. 10660951 Shop All Cytiva Products
Click to view available options
Diameter (Metric) Outer:
200 mm
300 mm
400 mm
600 mm
700 mm
1000 mm
Diameter (Metric) Inner:
16 mm
26 mm
50 mm
Height (Metric):
200 mm
300 mm
400 mm
600 mm
700 mm
1000 mm
Unit Size:
Each
Product Code. 10660951
Brand: Cytiva 18100082
Only null left
Add to basket
Edge
Add to basket
Specifications
For Use With (Equipment) Thermostat
Material Acrylic Plastic
Type Thermostat Jacket
Max. Temperature (Metric) 60°C
Height (Metric) 200 mm
Diameter (Metric) Inner 26 mm
Diameter (Metric) Outer 200 mm