Learn More
Acrylamide-1-13C, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Acrylamide-13 C |
| CAS | 287399-24-0 |
| Molecular Formula | 13 CC2H5NO |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i3+1 |
| SMILES | N[13 C](=O)C=C |
| IUPAC Name | (1 ^{13}C)prop-2-enamide |
| Molecular Weight (g/mol) | 72.07 |
| Formula Weight | 72.0405 g/mol |
| Percent Purity | 99 atom % 13 C, min 98% Chemical Purity |