Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Product Code. 17134611
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.65/0.2 μm
1.2 μm
Volume (Metric):
350 μL
1 mL
2 mL
Material (Membrane):
PTFE
PTFE (Hydrophilic)
Supor (PES)
Unit Size:
Pack of 10
Pack of 5
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Volume (Metric) Material (Membrane) unitSize
17134611 0.2 μm 350 μL PTFE Pack of 10
17114621 0.45 μm 1 mL Supor (PES) Pack of 5
17124621 1.2 μm 1 mL Supor (PES) Pack of 5
17144611 0.45 μm 350 μL PTFE Pack of 10
17174631 0.2 μm 2 mL PTFE Pack of 5
17164621 0.2 μm 1 mL PTFE Pack of 5
17174621 0.45 μm 1 mL PTFE Pack of 5
17104641 0.2 μm 350 μL PTFE (Hydrophilic) Pack of 10
17144641 0.2 μm 1 mL PTFE (Hydrophilic) Pack of 5
17154641 0.45 μm 1 mL PTFE (Hydrophilic) Pack of 5
17114641 0.45 μm 350 μL PTFE (Hydrophilic) Pack of 10
17184631 0.45 μm 2 mL PTFE Pack of 5
17184641 0.65/0.2 μm 2 mL PTFE (Hydrophilic) Pack of 5
17194641 0.45 μm 2 mL PTFE (Hydrophilic) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17134611 Supplier Cytiva Supplier No. 8047
Only null left
Add to basket
Add to basket
Optimized for a variety of applications requiring larger per well volume processing.
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Spezifikation
Material (Membrane) PTFE
Pore Size 0.2 μm
Sterility Non-sterile
For Use With (Application) Solvent Filtration
Quantity 10/Pk.
Volume (Metric) 350 μL