Learn More
Cytiva (formerly Pall Lab) AcroPak™ 800 Capsules with Fluorodyne™ II Membrane
AcroPak 800 Capsules with Fluorodyne II (PVDF) membrane that is compatible with aqueous and many organic solvents.
218.00€ - 259.00€
Specifications
Material (Membrane) | Hydrophilic PVDF |
---|---|
Sterility | Sterile |
Filtration Area | 800 cm2 |
Quantity | 1/Ea. |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
17154241
|
Cytiva (formerly Pall Lab)
12473 |
0.1 μm |
259.00€
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
17144241
|
Cytiva (formerly Pall Lab)
12471 |
0.2 μm |
218.00€
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
Description
- Double layer sterilizing membrane enhances reliability
- Upstream air vent prevents vapor lock
- Manufactured without the use of adhesives to minimize extractables
- Pre-sterilized by gamma irradiation
Specifications
Hydrophilic PVDF | |
Sterile | |
800 cm2 | |
1/Ea. |