Learn More
Cytiva AcroPak™ 500 Capsules with Supor™ Membrane, Sterile
Click to view available options
Pore Size:
0.1/0.1 μm
0.2/0.2 μm
0.8/0.2 μm
0.8/0.45 μm
Unit Size:
Each
Description
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Specifications
| Material (Membrane) | Hydrophilic PES |
| Pore Size | 0.1/0.1 μm |
| Sterility | Sterile |
| Filtration Area | 500 cm2 |
| No. per Pack | 1/Ea. |
| Outlet Style | Stepped Hose Barb |