Learn More
Cytiva AcroCap™ Positive Pressure Devices
Product Code. 17164321
Click to view available options
Pore Size:
0.1 μm
0.2 μm
0.45 μm
Unit Size:
Pack of 10
Product Code. 17164321
Brand: Cytiva 4482
Designed for fast, positive-pressure filtration of aqueous solutions up to 3 L.
  • Sterilization by gamma irradiation
  • Eliminates the risk of cytotoxic residuals from EtO sterilization
  • Integral hydrophobic vent prevents air lock
  • Product not approved for use in medical clinical surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) PES (Hydrophilic)
Pore Size 0.45 μm
Sterility Sterile
Filtration Area 15 cm
Inlet Style 1/4 in. hose barb inner taper
Outlet Style 1/4 in. hose barb inner taper
Quantity 10/Pk.