Learn More
Decon™ Neutralizing Acid Rinse
Click to view available options
Volume (Metric):
5 L
Unit Size:
Each
Description
- Suitable for glassware, ceramics, plastic, rubber, Stainless Steel and ferrous metals
- Bactericidal
Specifications
Specifications
| Product Type | Neutralizing Acid Rinse |
| Form | Liquid |
| Volume (Metric) | 5 L |
Safety and Handling
Product Identifier
- decon acid rinse
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : alcohols, C8-C18, ethoxylated propoxylated