Learn More
Decon™ Neutralizing Acid Rinse
Product Code. 11762098
Click to view available options
Volume (Metric):
5 L
Unit Size:
Each
Product Code. 11762098
Brand: Decon™ ACI5
Biodegradable. Decon Laboratories™ Neutraliziing Acid Rinse is non-foaming and rinses completely.
  • Suitable for glassware, ceramics, plastic, rubber, Stainless Steel and ferrous metals
  • Bactericidal
Specifications
Product Type Neutralizing Acid Rinse
Form Liquid
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • decon acid rinse
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : alcohols, C8-C18, ethoxylated propoxylated