Learn More
Olympus™ Achromatic Objective, SZX-ACH 1X
Product Code. 12550617
Change view
Click to view available options
For Use With (Equipment):
SZX microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12550617 SZX microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12550617 Supplier Olympus™ Supplier No. K10017142
  • ONLINE EXCLUSIVE PRICE
    795.00€
    433.28€ / Pack of 1
    Save 361.72€
    45% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Replace old or worn your objectives with quality Olympus parts. Olympus™ Achromatic Objective, SZX-ACH 1X fits SZX-series microscopes.
  • Achromatic
Specifications
Type Achromat Objective
Diameter (Metric) WD 90mm
For Use With (Equipment) SZX microscopes
Magnification Power 1X
Numerical Aperture 0.1