missing translation for 'onlineSavingsMsg'
Learn More

Acetyldiethylstilbestrol 2,3,4-Tri-O-acetyl-Beta-D-glucuronide Methyl Ester, TRC

Product Code. 30327282
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327282 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327282 Supplier Toronto Research Chemicals Supplier No. TRCA1736501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetyldiethylstilbestrol 2,3,4-Tri-O-acetyl-beta-D-glucuronide Methyl Ester
CAS 40269-22-5
Molecular Formula C33 H38 O12
Quantity 1 mg
Synonym Acetyldiethylstilbestrol 2,3,4-Tri-O-Acetyl-B-D-Glucuronide Methyl Ester
InChI Formula InChI=1S/C33H38O12/c1-8-26(22-10-14-24(15-11-22)40-18(3)34)27(9-2)23-12-16-25(17-13-23)44-33-31(43-21(6)37)29(42-20(5)36)28(41-19(4)35)30(45-33)32(38)39-7/h10-17,28-31,33H,8-9H2,1-7H3/b27-26+/t28-,29-,30-,31+,33+/m0/s1
SMILES CC\C(=C(\CC)/c1ccc(OC(=O)C)cc1)\c2ccc(O[C@@H]3O[C@@H]([C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]3OC(=O)C)C(=O)OC)cc2
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-[4-[(E)-4-(4-acetyloxyphenyl)hex-3-en-3-yl]phenoxy]oxane-2-carboxylate
Molecular Weight (g/mol) 626.65
Formula Weight 626.2363
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.