Learn More
Acetylcholine-d4 Chloride, TRC
Product Code. 30309157
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309157 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309157 Supplier Toronto Research Chemicals Supplier No. TRCA1720631MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Acetylcholine-d4 Chloride
CAS 344298-94-8
Molecular Formula C7 D4 H12 N O2 . Cl
Quantity 1 mg
Synonym Ethan-1,1,2,2-d4-aminium, 2-(acetyloxy)-N,N,N-trimethyl-, chloride (1:1) (ACI), 2-(Acetyloxy)-N,N,N-trimethylethan-1,1,2,2-d4-aminium chloride (1:1) (ACI), Ethan-1,1,2,2-d4-aminium, 2-(acetyloxy)-N,N,N-trimethyl-, chloride (9CI), Acetylcholine-1,1,2,2-d4 Chloride, Acetylcholine-d4 Chloride, Acetylcholine-1,1,2,2-D4 Chloride
InChI Formula InChI=1S/C7H16NO2.ClH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1/i5D2,6D2;
SMILES [Cl-].[2H]C([2H])(OC(=O)C)C([2H])([2H])[N+](C)(C)C
IUPAC Name (2-acetyloxy-1,1,2,2-tetradeuterioethyl)-trimethylazanium;chloride
Molecular Weight (g/mol) 185.685
Formula Weight 185.112