missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acetyl-Myelin Basic Protein (4-14)Peptide

Product Code. p-5384442
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418519 0.5 mg 0.5mg
15428519 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418519 Supplier Thermo Scientific™ Supplier No. J66946.!D

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Common Name Ac-MBP (4-14) Peptide
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 1432.70
Product Type Acetyl-Myelin Basic Protein (4-14) Peptide
Quantity 0.5 mg
Regulatory Status RUO
Sequence N-Acetyl-Gln-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.