Learn More
Acetonyltriphenylphosphonium bromide, 98+%
Product Code. 11430577
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430577 25 g 25g
11420577 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11430577 Supplier Thermo Scientific Alfa Aesar Supplier No. B22434.14
Request Bulk or Custom Format
CAS: 2236-01-3 | C21H20BrOP | 399.27 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2236-01-3
Molecular Formula C21H20BrOP
Molecular Weight (g/mol) 399.27
MDL Number MFCD00051863
InChI Key KZBWHCFTAAHIJJ-UHFFFAOYSA-M
Synonym acetonyltriphenylphosphonium bromide, acetonyl triphenylphosphonium bromide, 2-oxopropyl triphenylphosphanium bromide, phosphonium, 2-oxopropyl triphenyl-, bromide, acetonyltriphenylphosphoniumbromide, acmc-1cmpe, 2-oxopropyl-triphenylphosphonium bromide, 2-oxopropyl triphenylphosphonium bromide, 2-oxopropyl triphenyl phosphanium bromide, phosphonium, 2-oxopropyl triphenyl-,bromide 1:1
PubChem CID 638158
IUPAC Name 2-oxopropyl(triphenyl)phosphanium;bromide
SMILES [Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 226°C to 227°C
Quantity 25 g
Beilstein 3802148
Sensitivity Hygroscopic
Formula Weight 399.27
Percent Purity ≥98%
Chemical Name or Material Acetonyltriphenylphosphonium bromide