missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS/USP-Grade, 5, Pharmco

Product Code. 30286177
Change view
Click to view available options
Packaging:
5 Gallon Metal Pail
5 Gallon Poly Pail
Unit Size:
18.93 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
30286177 5 Gallon Poly Pail 18.93 L
30286183 5 Gallon Metal Pail 18.93 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30286177 Supplier Pharmco Supplier No. 329000000PL05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-64-1

Specifications

Quantity 5 Gallon
Packaging 5 Gallon Poly Pail
Boiling Point 56°C
Grade Reagent
Chemical Name or Material Acetone, ACS/USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.