Learn More
Acetamiprid-d3 (N-methyl-d3) (mixture of isomers), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Acetamiprid D3 (N-methyl D3) |
| CAS | 1353869-35-8 |
| Molecular Formula | C10 2H3 H8 Cl N4 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C10H11ClN4/c1-8(14-7-12)15(2)6-9-3-4-10(11)13-5-9/h3-5 H,6H2,1-2H3/b14-8+/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])N(Cc1ccc(Cl)nc1)\C(=N\C#N)\C |
| IUPAC Name | N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-(trideuteriomethyl)ethanimidamide |
| Molecular Weight (g/mol) | 225.69 |
| Formula Weight | 225.0861 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |