Learn More
Cole-Parmer™ Acetal Straight-through Quick-Disconnect Hose Barb Insert
Click to view available options
For Use With:
Fits 0.25 in. Tubing I.D.
Unit Size:
Each
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Acetal |
| Overall Length (Metric) | 6.35 cm |
| For Use With | Fits 0.25 in. Tubing I.D. |
| Min. Operating Temperature (Metric) | 82°C |
| Packaging Quantity | 1 Pack |
| Max. Operating Temperature (Metric) | 82°C |
| Product Type | Hose Barb Insert |