Learn More
Fisher Scientific™ Accu500 Water System
Product Code. 15429699 Shop All Fisherbrand Products
Click to view available options
Type:
UV Ultrapure Water System
UV Ultrapure Water System w/ Dispenser
Ultrapure Water Purification System
RO Flow Rate:
10 L/hr.
15 L/hr.
15 L/min.
20 L/hr.
30 L/hr.
Unit Size:
Each
Product Code. 15429699
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Exceeds ASTM, CLSI, CAP, and ISO Type I water standards. Fisher Scientific™ Accu500 Water System is a versatile purification system that can be used in a number of ways.
  • The fully automated control system contains pre-rinse, a flush feature preventing RO membrane fouling, RO membrane and cartridge life monitoring as well as many other programs for convenient operation
  • The pre-rinsing program starts automatically once a new RO membrane is installed so there is no need for user to set additional cleaning procedures
  • Two sets of dual-column purification cartridges work sequentially to ensure thorough removal of ions and organics in water
  • Automatic hourly system recirculation prevents bacterial growth
  • Resistivity is measured by a highly sensitive resistivity sensor
  • Electrical and water compartments are completely segregated, thus safer to operate
  • User can set water dispensing time for easy water collection
  • Both RO and ultrapure water can be dispensed from the water system
  • The system can be fitted with a 30L or 60L PE tank with a liquid level sensor and a tank vent filter with CO2 scavenger to protect purified water from atmospheric degradation
  • A 0.2μm filter or ultrafiltration filter (optional) removes final contaminants
  • Independent ultrapure water dispenser
  • The remote dispenser has a built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
  • Volumetric dispensing and calibration
  • The handle is 360 degree-rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility
Applications:
Ultrapure and RO water can be used in many areas. Some typical applications include:
Ultrapure water
  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.
RO water
  • Glassware cleaning
  • Washing machine for glassware
  • Waterbath water
  • Autoclaving
Specifications
Conductivity <20μS/cm
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Water Type Tap Water
Resistivity 18.2 Mohm.cm
Type Ultrapure Water Purification System
Feedwater Pressure 1 to 6 bar
Model Accu500
Particles >0.2µM/mL <1/mL with 0.2μm final filter or terminal ultrafiltration cartridge
Purified Water Flowrate Field 1.5 L/min.
RO Flow Rate 15 L/hr.
Temperature Range (Metric) 5°C to 45°C
Length (Metric) 30 cm
Height (Metric) 48 cm
Depth (Metric) 51 cm