Learn More
Miele Professional™ Accessory large washer-disinfector
Shop All Miele Products
Click to view available options
:
holder; for trays
Description
- For use in trays with 5 mm mesh
- 50 items per bag
Specifications
Specifications
| Material | Plastic |
| Product Type | Holder |
| Color | Grey |
| For Use With (Equipment) | Washer-disinfectors, medical |