missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Max. diameter 120 mm, max. height 470 mm
- For use with separately available injector module A 300/2
- For use with separately available injector module A 300/3
- For use with separately available injector module A 620
- Part of the easy load system
Specifications
Specifications
| Material | Stainless Steel, Plastic |
| Product Type | Injector Nozzle |
| Diameter (Metric) | 120 mm |
| Height (Metric) | 470 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?