Learn More
Miele Professional™ Accessory Booster Pump PLW8636 APLW 881
Shop All Miele Products
Click to view available options
Rack Type:
Rec out
Unit Size:
1 sheet
Description
- Rec in/out water inlet and drain for PLW 8636
- Required in order to pump a medium out of a buffer container
- For use with an existing on-site recycling tank
- Can be used with existing on-site container, e.g. IBC tank
- Installed by Customer Service
Specifications
Specifications
| Product Type | Booster Pump |
| Rack Type | Rec out |
| For Use With (Equipment) | Washer-Disinfectors |
| Quantity | 1 Ea. |