Learn More
Thermo Scientific™ Accessories for Eutech™ 6+ Conductivity/TDS/Salinity Meters
Product Code. 17868238
Change view
Click to view available options
Type:
Electrode Holder
Replacement Electrode
Unit Size:
Each
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17868238 Replacement Electrode Pack of 40
15347073 Electrode Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17868238 Supplier Thermo Scientific™ Supplier No. CONSEN91B
  • ONLINE EXCLUSIVE PRICE
    259.00€
    222.00€ / Pack of 40
    Save 37.00€
    14% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement electrodes and Grip-Clip™ electrode holder.
TRUSTED_SUSTAINABILITY
Specifications
Description Replacement electrode with ATC, k = 1.0
Type Replacement Electrode