Learn More
Thermo Scientific™ Accessories for MaxQ™ HP Orbital Shakers
Product Code. 12602795 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
7 Day Recorder, 220V
Gassing Manifold (One Gas)
Magnet and Pen Kit
Platform Adapter
Shaker Kit with Mount and Stand
For Use With (Equipment):
MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
MaxQ HP 416, 430, 420
MaxQ HP 416, 430, 435, 436, 480, 481
MaxQ HP 420
MaxQ HP 420, 435, 436, 480, 481
MaxQ HP 435, 436, 480, 481
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) unitSize
12602795 Platform Adapter MaxQ HP 416, 430, 435, 436, 480, 481 Each
12652785 Gassing Manifold (One Gas) MaxQ HP 420, 435, 436, 480, 481 Each
12632795 Magnet and Pen Kit MaxQ HP 435, 436, 480, 481 Each
12612795 Magnet and Pen Kit MaxQ HP 416, 430, 420 Each
10474432 Shaker Kit with Mount and Stand MaxQ HP 420 Each
10719273 7 Day Recorder, 220V MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12602795 Supplier Thermo Scientific™ Supplier No. 194040
Get maximum flexibility, regardless of your application, by using these accessories for Thermo Scientific™ MaxQ™ High Performance orbital shakers.
Get maximum flexibility, regardless of your application, by using Thermo Scientific™ Accessories for MaxQ™ High Performance Orbital Shakers.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Platform Adapter
Description Quick change platform adapter
For Use With (Equipment) MaxQ HP 416, 430, 435, 436, 480, 481