Learn More
Cytiva Accessories For Multiphor™ II Electrophoresis Systems
Product Code. 10279384 Shop All Cytiva Products
Click to view available options
Type:
Mylar Sheet
Sample Application Piece
Unit Size:
Pack of 200
Pack of 50
Product Code. 10279384
Brand: Cytiva 80112946
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Accessories for analytical or preparative separations of proteins and peptides by isoelectric focusing (IEF), native or SDS PAGE, 2-D PAGE and DNA electrophoresis
  • Assists high quality production of lab-cast gels
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
Type Sample Application Piece
Quantity 200/Pk