Learn More
Brady™ Accessories for BMP™21 Label Printers
Product Code. p-4826130
Click to view available options
Product Type:
Hard Case
Magnet
Rechargeable Li-ION Battery
Unit Size:
Each
Product Code. 11503864
Brand: Brady™ 110891
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Accessories for use with BMP21 portable label printers
  • AC Adapter: Convenient accessory for powering the BMP-21 Portable Label Printer without “AA” batteries
  • Hardside Carrying Case: Protects the label printer when transporting
  • Magnet Only Accessory: Allows you to attach label printer to panels, I-beams and any other magnetic-receptive surface during use or storage
  • Multifunctional Accessory: With additional options for carrying and operating the labeler, and a built-in flashlight for creating labels in dark spaces, this multifunctional tool is the ideal solution for fast, efficient labeling
Specifications
Product Type Magnet
For Use With (Equipment) BMP21 Label Printer
Width (English) 5.2 in.
Height (English) 4.1 in.
Height (Metric) 10.41 cm
Quantity 1 Ea.