Learn More
Thermo Scientific™ Accessories for BioCane™ Canister and Cane Systems
Use these accessories with Thermo Scientific™ BioCane™ Canister and Cane Systems.
Brand: Thermo Scientific™ AY509X12
Additional Details : Weight : 1.81000kg
Description
Use these accessories with Thermo Scientific™ BioCane™ Canister and Cane Systems.
- Warranty varies by country, please contact us for details
Specifications
Cryogenic Vial, 2.0mL; Case of 500 | |
7.06 cu. ft. | |
For use with Thermo Scientific BioCane canister and cane systems. |
Cryo Vial | |
2.0 mL |