Learn More
SymLine™ System Accessories
Click to view available options
Product Type:
Blind Plug
Cabinet Feed-through
Grounding Cable
Mounting Bracket
Color:
Black
Silver
Yellow
For Use With (Equipment):
32 mm Dia. Pipe
Cabinet (28 mm Borehole, 120 mm Wall Thickness)
Pipe System
SCAT SafetySystem Funnels
SCAT™ FlexTubes
Solvent System
Unit Size:
Each
Description
- SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
- All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
| Color | Silver |
| Diameter (Metric) Opening | 32 mm |
| Includes | Pipe Mounting |
| For Use With (Application) | SymLine™ System Customization |
| For Use With (Equipment) | Pipe System |
| Material | Metal |
| Product Type | Mounting Bracket |