Learn More
BINDER™ Access Port with Silicone Plug for CBF 170 CO2 Incubators
Shop All BINDER Products
Click to view available options
Diameter (Metric):
30 mm
Unit Size:
Each
Description
- Access port with silicone plug
- 30 mm inner diameter
- Right position
Specifications
Specifications
| Description | Right Position |
| Model | BINDER™ CBF 170 CO2 Incubators with Hot Air Sterilization and Humidity Regulation |
| Diameter (Metric) | 30 mm |