missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ac-rC Phosphoramidite, standard grade, serum vial bottle

Product Code. 17987162 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17987162 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17987162 Supplier Thermo Scientific™ Supplier No. 27140507
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Ac-rC CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis.

Ac-rC CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N4-acetyl-2'-O-TBDMSi-cytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: 5C47H64N5O9PSi
Base: 5rC
Base Protection: 5Ac
Formula weight: 5902.1 g/mol
CAS number: 5121058-88-6
HPLC Purity: 5≥98.0%
31P NMR Purity: 5≥98.0%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type Ac-rC CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Cytidine (C)
Reagent Features Fast deprotection
Shipping Condition Room Temperature
Purity 98%
Quantity 2 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.