Learn More
Ac-D-Trp-OH, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 2280-01-5 |
| Molecular Formula | C13H14N2O3 |
| MDL Number | MFCD00065021 |
| Quantity | 10 g |
| SMILES | O=C(O)[C@H](NC(C)=O)CC1=CNC2=C1C=CC=C2 |
| Molecular Weight (g/mol) | 246.26 |
| Percent Purity | 99% |
| Chemical Name or Material | Ac-D-Trp-OH |