Learn More
Ac-Asp-Glu-Val-Asp-7-Amino-4-TriFluoromethylcoumarin, MP Biomedicals™
Product Code. 11429895 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429895 10 mg 10mg
11419895 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11429895 Supplier MP Biomedicals Supplier No. 03AFC13810
  • ONLINE EXCLUSIVE PRICE
    784.00€
    746.55€ / 10mg
    Save 37.45€
    4% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Ac-Asp-Glu-Val-Asp-7-Amino-4-TriFluoromethylcoumarin is a fluorescent substrate.
  • Ac-DEVD-AFC is a sensitive fluorescent substrate for the assay of caspase-3 and CPP-32 activity.
  • The trifluoromethyl substituent of the fluorophore improves the membrane permeability of the substrate.
  • Protein or Enzyme Type: Activators, Inhibitors and Substrates
TRUSTED_SUSTAINABILITY
Specifications
Form Lyophilized
Quantity 10 mg
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.