Learn More
Tocris Bioscience™ AC 710
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AC 710 is a potent PDGFR family inhibitor (IC50 values are 1.2, 2, 7.7 and 10.5 nM for KIT, FLT3, PDGFRβ and CSF1R respectively; Kd = 1.3 nM for PDGFRα). Exhibits >30-fold selectivity over a panel of other kinases and exhibits no activity against cytochrome P450. Causes tumor regression of leukemia cell xenografts in mice. Also reduces joint swelling and inflammation in a mouse model of collagen-induced arthritis. Orally active.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | AC 710 |
| Purity | >99% |
| Molecular Weight (g/mol) | 562.7 |