Learn More
AC 220, TRC
Specifications
Specifications
| Chemical Name or Material | AC 220 |
| CAS | 950769-58-1 |
| Molecular Formula | C29H32N6O4S |
| Quantity | 10 mg |
| Synonym | AC 010220, Quizartinib, N-(5-tert-Butylisoxazol-3-yl)-N'-[4-[7-[2-(morpholin-4-yl)ethoxy]imidazo[2,1-b][1,3]benzothiazol-2-yl]phenyl]urea |
| InChI Formula | InChI=1S/C29H32N6O4S/c1-29(2,3)25-17-26(33-39-25)32-27(36)30-20-6-4-19(5-7-20)22-18-35-23-9-8-21(16-24(23)40-28(35)31-22)38-15-12-34-10-13-37-14-11-34/h4-9,16-18H,10-15H2,1-3H3,(H2,30,32,33,36) |
| SMILES | CC(C)(C)C1=CC(=NC(=O)NC2=CC=C(C=C2)C3=CN4C5=C(C=C(C=C5)OCCN6CCOCC6)SC4=N3)NO1 |
| IUPAC Name | 1-(5-tert-butyl-1,2-oxazol-3-yl)-3-[4-[6-(2-morpholin-4-ylethoxy)imidazo[2,1-b][1,3]benzothiazol-2-yl]phenyl]urea |
| Molecular Weight (g/mol) | 560.67 |