Learn More
Haleco™ NeutrAcide Gelling Powder for Acids
Product Code. 13277049
Change view
Click to view available options
Packaging Quantity:
9.5 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
13277049 9.5 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13277049 Supplier Haleco™ Supplier No. JLN120B
Absorbs and instantly solidifies all acid discharges, even hydrofluoric, regardless of concentration.
  • Product changes color from yellow to red, before returning to its initial color when the neutralization has finished
  • Solidification of spills facilitates cleaning
  • Non-toxic and incinerable
  • Used in the chemical industry, laboratories and for battery acids
  • Supplied in a 9.5kg bucket
Specifications
Material Sodium Carbonate, Cellulose
Type Gelling Powder
For Use With (Application) Absorption and solidification of acids in the laboratory and chemical industry
Packaging Quantity 9.5 kg
Weight (Metric) 9.5 kg