Learn More
ABsolute ABsolute™ qPCR Master Mixes, SYBR Green-Based Detection
Thermo Scientific™ ABsolute qPCR Master Mixes offer sensitivity and reproducibility for real-time PCR applications.
Brand: Thermo Scientific™ AB-1163/B
Additional Details : CAS : 7732-18-5 Weight : 0.80000kg
Description
Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ SYBR Green-Based Detection qPCR Master Mixes.
Highlights
- High Sensitivity & Specificity -The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
- Optimised buffer containing SYBR Green I
- Reproducibility -All master mixes undergo extensive QC to ensure batch to batch consistency
- Broad dynamic range
Specifications
ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, SYBR Green I, ROX™ passive reference dye (only in ROX mixes), Fluorescein (only in Fluorescein mix)); Separate MgCl2; Separate ROX vial (only in kits indicated). Store at -20°C - Avoid freeze/thaw cycles | |
ABsolute QPCR SYBR Green ROX (500nM) Mix | |
2000 x 50µL rxns | |
heated to 95°C for 15 minutes |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.