Learn More
BD AbSeq Enhancer Kit
Product Code. 32188681 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
AbSeq Enhancer Kit
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
32188681 AbSeq Enhancer Kit 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32188681 Supplier BD Supplier No. 570750
 Restricted Item
Estimated Shipment: 19-11-2026
Improves BD™ AbSeq performance by diminishing undesired Ab-Oligo background noise that may occur in certain applications.
This product should be used in workflows where samples are preserved in BD™ OMICS-Guard Sample Preservation Buffer prior to BD™ AbSeq staining, such as with BD™ Intracellular AbSeq. BD™ AbSeq Enhancer is a combination of 3 individual components that should be added in equal amounts to cells before they are stained with BD™ AbSeq.
TRUSTED_SUSTAINABILITY
Specifications
Product Type AbSeq Enhancer Kit