Learn More
Novus Biologicals™ AbSelect™ Serum Antibody Purification Kit
Fast and simple method to purify antibodies from ascites fluid and serum
662.89€ - 1153.32€
Specifications
Product Type Antibody Purification Kit
Content And Storage 4°C. Do not freeze.
View More Specs
Products 2
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
18086455
View Documents Promotion Details
Novus Biologicals™
863-0030
1 Purification
662.89€
1 test
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
18797083
View Documents Promotion Details
Novus Biologicals™
863-0500
3 Purifications
1153.32€
3 tests
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2024
Description
AbSelect Serum is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from ascites fluid and serum. The method involves the capture of the antibody on the AbSelect Serum resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
  •  Compatible with the lightning-link antibody labeling kits
  • Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye
Specifications
Specifications
Antibody Purification Kit
4°C. Do not freeze.