Learn More
Novus Biologicals™ AbSelect™ Serum Antibody Purification Kit
Fast and simple method to purify antibodies from ascites fluid and serum
662.89€ - 1153.32€
Specifications
Product Type | Antibody Purification Kit |
---|---|
Content And Storage | 4°C. Do not freeze. |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
18086455
|
Novus Biologicals™
863-0030 |
1 Purification |
662.89€
1 test |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
18797083
|
Novus Biologicals™
863-0500 |
3 Purifications |
1153.32€
3 tests |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
Description
AbSelect Serum is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from ascites fluid and serum. The method involves the capture of the antibody on the AbSelect Serum resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
- Compatible with the lightning-link antibody labeling kits
- Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye
Specifications
Antibody Purification Kit | |
4°C. Do not freeze. |