Learn More
Novus Biologicals™ AbSelect™ Serum Antibody Purification Kit
Product Code. 18797083 Shop All Bio Techne Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
18797083 3 Purifications 3 tests
18086455 1 Purification 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18797083 Supplier Novus Biologicals™ Supplier No. 8630500
 Restricted Item
Estimated Shipment: 23-10-2026
Fast and simple method to purify antibodies from ascites fluid and serum
AbSelect Serum is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from ascites fluid and serum. The method involves the capture of the antibody on the AbSelect Serum resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
  •  Compatible with the lightning-link antibody labeling kits
  • Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye
Recommended Storage
Store at 4°C. Do not freeze.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibody Purification Kit
Sufficient For 3 Purifications
Content And Storage 4°C. Do not freeze.
Quantity 3 Tests