Learn More
Novus Biologicals™ AbSelect™ G Antibody Purification Kit
Product Code. p-6376305 Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
Product Code. 18051254
Brand: Novus Biologicals™ 8900005
For purification of antibodies from crude samples such as ascites fluid or immune serum and removal of contaminants
The method involves capturing the antibody on the AbSelect Protein G resin. Protein G has a high affinity for the Fc regions of IgG molecules from a variety of species and once the antibody has bound to the Protein G, unwanted substances can be removed by simply washing the resin. The purified product is then eluted and neutralized.
  •  Compatible with the lightning-link and lightning-link rapid conjugation systems
    • Allows purified antibody to be immediately labeled with a hands-on time of under 30 seconds
Recommended Storage
Store at 4°C. Do not freeze.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibody Purification Kit
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test