Learn More
Novus Biologicals™ AbSelect™ G Antibody Purification Kit
Product Code. 18051254 Shop All Bio Techne Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
18051254 1 Purification 1 test
18029393 3 Purifications 3 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18051254 Supplier Novus Biologicals™ Supplier No. 8900005
 Restricted Item
Estimated Shipment: 22-10-2026
For purification of antibodies from crude samples such as ascites fluid or immune serum and removal of contaminants
The method involves capturing the antibody on the AbSelect Protein G resin. Protein G has a high affinity for the Fc regions of IgG molecules from a variety of species and once the antibody has bound to the Protein G, unwanted substances can be removed by simply washing the resin. The purified product is then eluted and neutralized.
  •  Compatible with the lightning-link and lightning-link rapid conjugation systems
    • Allows purified antibody to be immediately labeled with a hands-on time of under 30 seconds
Recommended Storage
Store at 4°C. Do not freeze.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibody Purification Kit
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test