Learn More
Testo™ 110 Single Channel NTC Thermometer
Product Code. 10324731
Click to view available options
Temperature Range (Metric):
-50°C to +150°C
Unit Size:
Each
Product Code. 10324731
Brand: Testo™ 0560 1108
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Measure temperatures in very low temperature environments. Testo™ 110 Single Channel NTC Thermometers are ideal for use in cold storage rooms, cold storage warehouses and the outdoors.
  • 1-Channel NTC Thermometer
  • Extremely accurate, also in very low temperature range such as cold storage rooms
  • Optional connectable temperature sensor and wireless sensor for different applications in the field
  • Audible alarm, user-defined thresholds
  • HACCP compliant, certified according to EN 13485 for use in the food sector
Specifications
Material ABS Plastic
Max. Temperature (Metric) 150°C
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +150°C
Accuracy ±0.2°C
Width (Metric) 64 mm