Learn More
Esculape Laboratoire™ ABS First AID Kits
Click to view available options
Depth (Metric):
17 cm
20 cm
34 cm
Unit Size:
Each
Specifications
Specifications
| Depth (Metric) | 34 cm |
| Length (Metric) | 36 cm |
| Product Type | First Aid Kit |
| Color | Blue |
| Material | ABS |
| Includes | 4 Gas band 3m x 7cm, 4 Extensible web 3m x 7cm, 4 Anti-compress shots, 20 Sterile compresses 20 x 20, Survival blanket, Doigtier, 12 Safety pin, Guide first aid, Scissors, 2 Pair of Vinyl gloves, 2 Sterile pressure dressing 2.5m x 10cm, 2 sterile pressure |
| Quantity | 1 Ea. |
| Height (Metric) | 12 cm |