Learn More
Esculape Laboratoire™ ABS First AID Kits
Product Code. 11813232
Change view
Click to view available options
Depth (Metric):
17 cm
20 cm
34 cm
Unit Size:
Each
Product Code. Depth (Metric) unitSize
11813232 20 cm Each
11823232 34 cm Each
11803232 17 cm Each
3 options
Product Code. 11813232
Brand: Esculape Laboratoire™ 720 01 02
Only null left
Specifications
Depth (Metric) 20 cm
Length (Metric) 26 cm
Product Type First Aid Kit
Color Blue
Material ABS
Includes 2 Gas band 3m x 7cm, 2 Extensible web 3m x 7cm, 3 Anti-compress shots, 10 Sterile compresses 20 x 20, Doigtier, Guide first aid, 6 number of clips, Scissors, 2 Pair of Vinyl gloves, Sterile pressure dressing 2.5m x 10cm, Sterile pressure dressing 2.5 x 6c
Quantity 1 Ea.
Height (Metric) 8.5 cm