missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fluorescent Dye 749-C7 bisAcid

Product Code. 16294880
Change view
Click to view available options
:
Quantity: 5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16294880 Quantity: 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16294880 Supplier Abnova™ Supplier No. U0223.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cyanine 7 dyes are common near infrared red fluorophores used in in vivo imaging applications

Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis

  • Chemical Formula: C40H47F3N2O12S2
  • Mol. Weight: 868.93g/mol
  • Purity: ≥90%
  • Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
  • Excitation (Max): 749nm
  • Emission (Max): 776nm
  • Solubility: DMSO, water

Application: Conjugation, In vivo imaging

Recommended Storage: Store in the dark at -20°C.

Specifications

Description Cyanine 7 dyes are common near infrared red fluorophores used in in vivo imaging applications.
For Use With (Application) Conjugation; in vivo Imaging
Quantity 5 mg
Product Type Fluorescent Dye 749-C7 bisAcid
Molecular Weight (g/mol) 868.93g/mol
Purity ≥90%
Content And Storage Store in the dark at -20°C
Label or Dye Fluorescent Dye 749-C7 bisAcid
Emission 776
Excitation 749

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.