Learn More
Abnova™ Fluorescent Dye 700-I Phalloidin
Description
In comparison with Alexa Fluor 700, Fluorescence Dye 700-I are brighter with stronger absorption at 633nm. Fluorescence emission of Fluorescence Dye 700-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.
- Spectral Equivalent: Alexa Fluor 700
- Molar Extinction Co-efficient: ≥2,00,000 M-1cm-1
- Excitation (Max): 685nm
- Emission (Max): 710nm
- Solubility: DMSO
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye 700-I with Phalloidin conjugate. In comparison with Alexa Fluor 700, Fluorescence Dye 700-I are brighter with stronger absorption at 633nm. Fluorescence emission of Fluorescence Dye 700-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 300 Reactions |
| Product Type | Fluorescent Dye 700-I Phalloidin |
| Sufficient For | 300 Reactions |
| Equivalence | Alexa Fluor 700 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 700-I Phalloidin |
| Emission | 710 |
| Show More |
For research use only (RUO).
Your input is important to us. Please complete this form to provide feedback related to the content on this product.