missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fluorescent Dye 610-I Succinimidyl Ester
Description
Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 610
- Purity: ≥90%
- Molar Extinction Co-efficient: >80,000 M-1cm-1
- Excitation (Max): 605nm
- Emission (Max): 627nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. |
| For Use With (Application) | Conjugation |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 610-I Succinimidyl Ester |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 610 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 610-I Succinimidyl Ester |
| Emission | 627 |
| Show More |
For research use only (RUO).
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?